For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.
Protein Form: Full Length
MARK2 is a serine/threonine protein kinase involved in the control of cell polarity and microtubule stability. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1. These isoforms were characterized by the presence of a 162-bp alternative exon that gave rise to two forms, one containing the exon and the other one lacking it. Both forms were found to be coexpressed in a number of selected cell lines and tissue samples. The human EMK1 was shown to be encoded by a single mRNA ubiquitously expressed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELKL motif kinase; ELKL motif kinase 1; EMK-1; MAP/microtubule affinity-regulating kinase 2; MARK2; MGC99619; OTTHUMP00000218338; OTTHUMP00000218339; OTTHUMP00000218340; OTTHUMP00000218343; Par-1b; PAR1 homolog; PAR1 homolog b; Ser/Thr protein kinase PAR-1B; serine/threonine protein kinase EMK; Serine/threonine-protein kinase MARK2; testicular tissue protein Li 117
Gene Aliases: EMK-1; EMK1; MARK2; PAR-1; Par-1b; Par1b
UniProt ID: (Human) Q7KZI7
Entrez Gene ID: (Human) 2011
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support