LanthaScreen™ Tb-labeled Anti-Goat Ab
Product Image
Thermo Scientific™
LanthaScreen™ Tb-labeled Anti-Goat Ab
A general reagent for use in assay development, this terbium labeled antibody will bind goat antibodies. This product is ideallyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV377025 μg
PV37721 mg
Catalog number PV3770
Price (USD)/ 25 µg
1,050.00
-
Add to cart
Quantity:
25 μg
Price (USD)/ 25 µg
1,050.00
Add to cart
LanthaScreen™ Tb-labeled Anti-Goat Ab
Catalog numberPV3770
Price (USD)/ 25 µg
1,050.00
-
Add to cart
A general reagent for use in assay development, this terbium labeled antibody will bind goat antibodies. This product is ideally suited for use during assay development when evaluating multiple primary goat antibodies or when you are unable to directly terbium label your primary antibody.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Excitation/Emission340/490, 520
Quantity25 μg
ReactivityGoat
ReadoutEnd Point
Shipping ConditionDry Ice
ConjugateLanthanide Chelates, Terbium (Tb)
Product LineLanthaScreen™
Unit Size25 µg
Contents & Storage
Terbium-labeled anti-Goat antibody in HEPES buffered saline. Store at -20°C.