Get accurate thickness and moisture data, improve productivity, and precisely control raw materials use with a full-spectrum infrared sensor for web producing applications.
Producers of plastic films, extrusion coatings, blown films, and nonwovens can get accurate thickness and moisture data, improve product quality, and precisely control raw material use with an easily-calibrated infrared gauge. Whether you need a coating thickness gauge, paint thickness gauge, or film thickness gauge, the Thermo Scientific™ PROSIS™ Infrared Process Analysis Thickness Sensor uses the full spectral response of the near infrared range to analyze materials on the line, providing precise, multi-component thickness data that improves production quality and reduces waste.
The PROSIS Infrared Process Analysis Thickness Sensor for web gauging simultaneously measures and analyzes the entire NIR spectrum instead of a few preselected filters that only give you a small part of the picture. The robust measurement performance of this infrared gauge delivers data quickly to increase yield, reduce scrap and raw material costs, and improve product quality.
Typical applications for Reflection PROSIS IR Sensor: