Start-to-finish LC-MS workflow solutions designed to increase productivity, reduce costs and address the ever-evolving demands for pesticide residue testing.
Catalog Number | Description | Mass Spectrometer Interface | Methods |
---|---|---|---|
PE-OTVF-01 | Triple Quadrupole Quantitation | Thermo Scientific™ TSQ Quantis™ Triple Quadrupole Mass Spectrometer | Quantitation method: > 400 pesticides (SRM) |
PE-OTVF-02 | HRAM Quantitation | Thermo Scientific™ Orbitrap Exploris™ 120 Mass Spectrometer | Quantitation method: > 500 pesticides (DIA or FSddMS2) |
PE-QVFB-03 | HRAM Screening and Quantitation | Thermo Scientific™ Orbitrap Exploris™ 120 Mass Spectrometer | Quantitation and Screening methods: > 500 pesticides or more (DIA or FSddMS2) |
Discover compelling productivity and efficiency enhancements for laboratories performing pesticide residue testing. Thermo Scientific™ Pesticide Explorer™ is a comprehensive liquid chromatography-mass spectrometry (LC-MS) solution designed for laboratories performing routine quantitation and pesticide residue screening in food matrices.
The solution includes all the workflow components needed—consumables, hardware, software and built-in instrument and data processing methods—pre-configured and tested, from your single, trusted supplier, Thermo Fisher Scientific.
Pesticide Explorer provides start-to-finish workflows tailored to help food monitoring and testing laboratories reduce startup time and cost. It provides compelling productivity and efficiency enhancements for both startup laboratories and laboratories adding new analytical capabilities to address evolving customer and industry demands. Regardless of staff or laboratory expertise, Pesticide Explorer is designed to make it easier for both new and experienced users to obtain reliable, unambiguous, high-quality LC-MS/MS results.