pHrodo™ AM Variety Pack
pHrodo™ AM Variety Pack
Invitrogen™

pHrodo™ AM Variety Pack

Measure and quantify intracellular pH with the pHrodo AM Variety Pack, which includes pHrodo Red and Green AM indicator dyes,Read more
Have Questions?
Catalog number P35380
Price (USD)/ 1 kit
258.65
Online exclusive
263.00 
Save 4.35 (2%)
-
Add to cart
Price (USD)/ 1 kit
258.65
Online exclusive
263.00 
Save 4.35 (2%)
Add to cart
pHrodo™ AM Variety Pack
Catalog numberP35380
Price (USD)/ 1 kit
258.65
Online exclusive
263.00 
Save 4.35 (2%)
-
Add to cart
Measure and quantify intracellular pH with the pHrodo AM Variety Pack, which includes pHrodo Red and Green AM indicator dyes, fluorogenic probes that traverse the cell membrane and remain within the intracellular space upon cleavage by nonspecific esterases.

The pHrodo AM Variety Pack contains one vial each of the pHrodo Red AM and pHrodo Green AM indicator dyes, with each vial providing enough reagent volume to test one 96-well plate. pHrodo Red AM and pHrodo Green AM intracellular pH indicators are fluorogenic probes used to measure intracellular pH in live cells. They are weakly fluorescent at neutral pH, but dramatically increase in brightness as the pH decreases. These reagents can quantify cellular cytosolic pH in the range of 4–9 with a pKa of ∼6.5 and have excitation/emission spectra of 560/585 nm (pHrodo Red) and 509/533 nm (pHrodo Green).

pHrodo Red AM and pHrodo Green AM dyes are photostable and appropriate for use in multiplexing experiments; pHrodo Red easily multiplexes with a wide variety of blue, green, and far-red dyes and reporters such as GFP, Fuo-4, calcein, NucBlue, CellEvent Caspase 3/7 green, Mitosox Green, and Mitotracker Deep Red, among many others. pHrodo Green dye easily multiplexes with a wide variety of blue, red, and far-red dyes reporters such as Mitosox Red, CellEvent Caspase 3/7 Red, NucBlue, RFPs, and Mitotracker Deep Red, among many others. The pHrodo Red and pHrodo Green dyes have been modified with AM ester groups, which results in an uncharged molecule that can permeate cell membranes. Once inside the cell, the lipophilic blocking groups are cleaved by nonspecific esterases, resulting in a compound that is retained within the intracellular space. Fluorescence intensity of the probe is then an indicator of intracellular pH. Subsequent use of the Intracellular pH Calibration Buffer Kit (Cat. No. P35379) allows this intracellular pH to be quantified.

pHrodo Red AM and pHrodo Green AM intracellular pH indicators are compatible with various platforms, such as traditional fluorescence microscopy, high content screening (HCS), flow cytometry, and microplate-based fluorimetry or high throughput screening (HTS). These reagents are also compatible with widefield or confocal fluorescence microscopes, flow cytometers, fluorescent plate readers, and high content instruments.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
Cell PermeabilityCell-Permeant
ColorGreen, Orange
DescriptionpHrodo™ AM Variety Pack
Detection MethodFluorescence
For Use With (Application)Cell Analysis
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Fluorometer, High Content Analysis Instrument
Product TypeFluorogenic Probe
EmissionVisible
FormLiquid
Format96-well Plate
IndicatorpH Indicator
Indicator PropertiesFluorescent Dye-Based
Product LinepHrodo™
Quantity1 Kit
SolubilityDMSO (Dimethylsulfoxide)
Sub Cellular LocalizationCytoplasm
Sufficient ForTesting one 96-well plate
Excitation/EmissionpHrodo™ Green: 509/533 nm
pHrodo™ Red: 560/585 nm
Unit Size1 kit
Contents & Storage
Store at 2–8°C, desiccate, and protect from light.