Perform fast, flexible, and efficient instrument loading for various high-throughput applications along with inventory management and sample tracking.
Catalog Number | Description |
---|---|
ORB2004 | Orbitor RS2 Bundle, 6 Stacks |
ORB2001 | Orbitor RS2 Bundle, 3 Hotels |
ORB2002 | Orbitor RS2 Bundle, 3 Stacks |
ORB2003 | Orbitor RS2 Bundle, 6 Hotels |
ORB2005 | Orbitor RS2 Mixed Bundle, any combination of Hotels and Stacks; 3 total |
ORB2006 | Orbitor RS2 Mixed Bundle, any combination of Hotels and Stacks; 6 total |
Increase throughput, storage capacity, and operational efficiency with the Thermo Scientific™ Orbitor™ RS2 Microplate Mover, an updated, compact, and collaborative bench-top mover with integrated barcode scanning for greater flexibility. The Orbitor RS2 microplate mover increases efficiency as the centerpiece of various high-throughput automation operations. Its innovative bi-directional telescoping arm provides exceptional reach and precision, while its expansive 360° workspace makes it a great laboratory productivity partner. This microplate robot is ideal for high-throughput applications and is available in a variety of configurations to match your needs.
Choose the configuration of hotels and stacks to match your requirements