The thiol reactive Oregon Green™ 488 maleimide can be used to can be used to create green fluorescent bioconjugates withRead more
Have Questions?
Catalog number O6034
Price (USD)/ 5 mg
384.65
Online exclusive
388.00
Save 3.35
-
Add to cart
Price (USD)/ 5 mg
384.65
Online exclusive
388.00
Save 3.35
Add to cart
Oregon Green™ 488 Maleimide
Catalog numberO6034
Price (USD)/ 5 mg
384.65
Online exclusive
388.00
Save 3.35
-
Add to cart
The thiol reactive Oregon Green™ 488 maleimide can be used to can be used to create green fluorescent bioconjugates with excitation/emission maxima ∼496/524 nm. This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∼ 4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity5 mg
Product TypeMaleimide
Chemical ReactivityThiol
Shipping ConditionRoom Temperature
Reactive MoietyMaleimide
Label or DyeOregon Green™ 488
Label TypeClassic Dyes
Product LineOregon Green™
Emission524
Excitation496
Unit Size5 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.