NuPAGE™ Bis-Tris Welcome Pack, 10%
NuPAGE™ Bis-Tris Welcome Pack, 10%
Invitrogen™

NuPAGE™ Bis-Tris Welcome Pack, 10%

The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the Mini Gel Tank.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
NP030A10-well
NP030B12-well
NP030C15-well
Catalog number NP030A
Price (USD)/ 1 kit
594.65
Online exclusive
614.00 
Save 19.35 (3%)
-
Add to cart
Wells:
10-well
Price (USD)/ 1 kit
594.65
Online exclusive
614.00 
Save 19.35 (3%)
Add to cart
NuPAGE™ Bis-Tris Welcome Pack, 10%
Catalog numberNP030A
Price (USD)/ 1 kit
594.65
Online exclusive
614.00 
Save 19.35 (3%)
-
Add to cart
The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

The NuPAGE Bis-Tris Welcome Pack contains:
• Mini Gel Tank (A25977)
• NuPAGE Bis-Tris mini gels (2 boxes, 20 gels)
• NuPAGE MES SDS Running Buffer, 20X (NP0002)
• NuPAGE LDS Sample Buffer, 4X (NP0007)
• NuPAGE Sample Reducing Agent, 10X (NP0009)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)

About the Mini Gel Tank
The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.

About NuPAGE Bis-Tris gels
Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use.

Transfer
For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Mini Gel Tank
Gel Percentage10%
Gel SizeMini
Gel Thickness1.0 mm
Gel TypeBis-Tris
Separation Range3.5 to 260 kDa
Wells10-well
Product LineNuPAGE™
Quantity1 Welcome Pk.
Shipping ConditionApproved for shipment at room temperature and wet ice
Unit Size1 kit