NuPAGE Sample Reducing Agent (10X) is used to reduce protein samples for protein gel electrophoresis. It contains 500 mM dithiothreitolRead more
Have Questions?
Change view
Catalog Number
Quantity
NP0009
10mL
NP0004
250μL
2 Options
Catalog number NP0009
Price (USD)/ 10 mL
75.65
Online exclusive
77.25
Save 1.60 (2%)
-
Add to cart
Quantity:
10mL
Price (USD)/ 10 mL
75.65
Online exclusive
77.25
Save 1.60 (2%)
Add to cart
NuPAGE™ Sample Reducing Agent (10X)
Catalog numberNP0009
Price (USD)/ 10 mL
75.65
Online exclusive
77.25
Save 1.60 (2%)
-
Add to cart
NuPAGE Sample Reducing Agent (10X) is used to reduce protein samples for protein gel electrophoresis. It contains 500 mM dithiothreitol (DTT) for a 10X concentration in a stabilized liquid form.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration500 mM
Product TypeSample Reducing Agent
Shelf Life6 Months
Product LineNuPAGE™
Quantity10mL
Shipping ConditionWet Ice
Unit Size10 mL
Contents & Storage
NuPAGE™ Sample Reducing Agent (500 mM dithiothreitol) should be stored at 4°C. Guaranteed stable for 6 months unless otherwise specified in the product literature.