MicroAmp® Cap Strips are designed to fit on MicroAmp® reaction tubes, tube strips, and 96-well plates. They are available inRead more
Have Questions?
Catalog number N8010835
Price (USD)/ 300 strips
130.65
Online exclusive
143.00
Save 12.35 (9%)
-
Add to cart
Price (USD)/ 300 strips
130.65
Online exclusive
143.00
Save 12.35 (9%)
Add to cart
MicroAmp™ 8-Cap Strip, assorted colors
Catalog numberN8010835
Price (USD)/ 300 strips
130.65
Online exclusive
143.00
Save 12.35 (9%)
-
Add to cart
MicroAmp® Cap Strips are designed to fit on MicroAmp® reaction tubes, tube strips, and 96-well plates. They are available in 8-cap and 12-cap strips in clear or an assorted set of red, orange, blue, and green. They have a domed profile and are suitable for end-point PCR. For real-time PCR applications, please see our MicroAmp® Optical 8-Cap Strips (cat. no. 4323032), which have a flat-top profile. Attach the caps using the MicroAmp® Cap Installing Tool.
• Will not interfere with sample reading • Provides tight seal to minimize evaporation • Easy to apply and remove
Minimize Evaporation Use MicroAmp® Caps Strips to seal the tubes containing your precious samples. This will reduce the possibility of contamination among sample wells and help ensure reliable results.
Easy to Use Use the MicroAmp® Cap Installing Tool to push the caps down and form a tight seal. When ready, just peel away the caps using the tabs at the end.
Colored for Organization These caps are available in different colors to help you keep track of your tubes during assay preparation.
Specifications
Certifications/ComplianceCertified DNA/Rnase Free
ColorAssorted
Volume (Metric)0.2 mL
Quantity300 strips
For Use With (Equipment)Thermal Cyclers
Product LineMicroAmp™
MaterialPolypropylene
Shipping ConditionRoom Temperature
Closure TypeDomed Caps
No. of Wells96
Format200 Strip Caps
Product Type8-Strip Cap
Unit Size300 strips
Contents & Storage
Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Assorted colors.