GeneAmp™ Thin-Walled Reaction Tube, with domed cap, 0.5 mL, autoclaved
Applied Biosystems™ GeneAmp™ Thin-Walled Reaction Tubes enable fast heat transfer for precise PCR in 0.5 mL PCR Systems.• Thin plasticRead more
Have Questions?
Catalog number N8010611
Price (USD)/ 1,000 tubes
271.00
-
Add to cart
Price (USD)/ 1,000 tubes
271.00
Add to cart
GeneAmp™ Thin-Walled Reaction Tube, with domed cap, 0.5 mL, autoclaved
Catalog numberN8010611
Price (USD)/ 1,000 tubes
271.00
-
Add to cart
Applied Biosystems™ GeneAmp™ Thin-Walled Reaction Tubes enable fast heat transfer for precise PCR in 0.5 mL PCR Systems. • Thin plastic enables fast heating and cooling for accurate results • Flat, frosted caps for easy marking • Captive lid with positive-click closure assures proper seating • Polished surface and conical bottom allow maximum sample recovery • Autoclaved tubes provide a clean, controlled start
Rapid Heat Transfer The GeneAmp™ Reaction Tube is made of specially engineered polypropylene and is optimized for use with the Veriti™ 60-Well Thermal Cycler and the Dual 96-Well GeneAmp™ PCR System 9700. Our thin-walled tubes facilitate rapid heat transfer to and from samples to achieve faster cycle times and increased specificity. Like all our products, GeneAmp™ Reaction Tubes undergo stringent quality testing to ensure reproducible results.
Easy to Use The GeneAmp™ Reaction Tube features design elements such as a flat, frosted cap, a tight seal, and a clear conical bottom that bring high performance to everyday applications. Recommended maximum reaction volume is 100 μL.
Note: See user's manual or package insert for limited label license and trademark information.
Specifications
Certifications/ComplianceCertified DNA/Rnase Free
ColorNatural
Volume (Metric) Working0.5 mL
Volume (Metric)0.6 mL
Quantity1000 tubes
For Use With (Equipment)Thermal Cyclers
Product LineGeneAmp™
SterilitySterile
MaterialPolypropylene
Reaction SpeedFast
Shipping ConditionRoom Temperature
Closure TypeDomed Caps
No. of Wells96
AutoclavableAutoclavable
Format1,000 Tubes
Product TypeReaction Tube
Unit Size1,000 tubes
Contents & Storage
Contains 1,000 GeneAmp™ Thin-Walled Reaction Tubes with Domed Cap, 0.5 ml, Autoclaved. Store at room temperature.