Specifications
ClassificationAnimal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum
Cell TypeStem Cells, Primary Cells, Hard-to-Transfect Cells
Format6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample TypePlasmid DNA, Synthetic siRNA
Transfection TechniqueElectroporation
For Use With (Equipment)Neon™ Transfection System
For Use With (Application)Transfection
No. of Reactions25 x 2
Product LineNeon™
Product TypeTransfection System
Quantity25 x 2 Reactions
Serum CompatibleYes
Shipping ConditionRoom Temperature
Starting Material Amount2x10^4 - 6 x10^8 cells; Varies by cell type
System TypeNeon™ Transfection System
Volume (Metric) Sample10 μl
Unit Size25 x 2 reactions