Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Actual product may vary
Thermo Scientific™
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Improve the functionality of Savant SpeedVac Concentrators with these Savant SpeedVac Concentrators miscellaneous accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
MF190Mist filter replacement cartridge
SCT120Chemical Trap (order cartridges separately)
DTK120RChemical Trap Kit for Radioactivity
DC120Disposable Cartridge (trapping volatile radioactivity)
DC120ADisposable Cartridge (acid neutralization)
ANC100Acid Neutralizing Compound (acetic acid)
UTP-TEF0.75
also known as UTP-TEF 0.75
PTFE tubing pack; For use with SPD140DDA SpeedVac Concentrators
ANS121Ammonia Neutralizing Solution (Oligo Prep)
ANT100Post Trap Assembly (for Oligo Prep; supplied with DNA130)
GCF400Glass condensation flask
CC120DX
also known as CC120/DX
Deluxe Cart
Catalog number MF190
Price (USD)/ Each
-
Request A Quote
Description:
Mist filter replacement cartridge
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
Specifications
DescriptionMist filter replacement cartridge
TypeMist Filter Replacement Cartridge
For Use With (Equipment)OME190 Oil Mist Eliminator for VLP Series Vacuum Pump
Unit SizeEach