Sensititre™ Gram Negative Susceptibility Testing Plate, IVD format
Sensititre™ Gram Negative Susceptibility Testing Plate, IVD format
Thermo Scientific™
Sensititre™ Gram Negative Susceptibility Testing Plate, IVD format
NEW antimicrobial options for difficult to treat Gram-negative infections in a high-quality, ready to use AST broth microdilution (BMD) plate format, approved by the FDA and meeting the latest CLSI breakpoints.
Have Questions?
Catalog number MDRGN2F
Price (USD)/ Pack of 10
-
Request A Quote
Description:
Sensititre Gram Negative AST Plate
Expand your Gram-negative organism susceptibility testing options by accessing the latest antibiotics including cefiderocol and imipenem/relebactam. The Thermo Scientific™ Sensititre™ MDRGN2F plate delivers results directly from broth microdilution (BMD), the gold standard1 phenotypic method for Antimicrobial Susceptibility Testing (AST). Open access to one of the most comprehensive range of second-line antimicrobials, on a single plate format, designed for in-vitro diagnostic use.
  • Updated standard format including Cefiderocol and Imipenem/Relebactam 
  • IVD-labelled 
  • BMD provides accurate MIC results guiding treatment optimization
  • Meets the latest CLSI breakpoints
  • Sensititre MIC determination is achieved using direct measurement rather than relying on extrapolated data, thereby providing an accurate MIC result, not an estimate. 
  • All contained antibiotics also available in customizable plate options, along with access to over 300 antimicrobials, to enable meeting local formulary and Stewardship program requirements
Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual test options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing.
The Sensititre System utilizes gold standard-level MIC accuracy1, providing superior quality and reproducibility for accurate results the first time. Broth microdilution is the reference method for AST, and in many cases 2 the most reliable phenotypic method to guide therapeutic decisions.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Antimicrobial AgentsVarious
DescriptionSensititre Gram Negative AST Plate
Antibiotic CodeVarious
Product TypeAST Plate
Target Organism ClassMultidrug-resistant, Gram-negative Organisms
Test TypeMicrobial Susceptibility
Quantity10/Pk.
Unit SizePack of 10