Specifications
Chemical Name or Material2-Propanol (45%), Acetonitrile (45%), and Acetone (10%) Solution
pH6 to 7
Name NoteMobile-phase solvent blend
Absorbance≤1.0 AU at 210nm, ≤1.0 AU at 220nm
ColorColorless
CAS67-64-1
CAS75-05-8
Density0.787 g/cm3
Boiling Point75°C
Quantity1 L
PackagingBorosilicate Glass Bottle
Physical FormLiquid
Synonymisopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Percent PurityPass Test (By GC-FID)
GradeLC-MS
Concentration or Composition (by Analyte or Components)45% 2-Propanol: 45% Acetonitrile: 10% Acetone
Unit SizeEach