Novex™ IEF Sample Buffer pH 3-7 (2X)
Product Image
Invitrogen™

Novex™ IEF Sample Buffer pH 3-7 (2X)

Novex® IEF Sample Buffer pH 3-7 (2X) is optimized pre-mixed IEF sample buffer for using with Novex® pH 3-7 IEFRead more
Have Questions?
Catalog number LC5371
Price (USD)/ 25 mL
27.65
Online exclusive
28.41 
Save(3%)
-
Add to cart
Price (USD)/ 25 mL
27.65
Online exclusive
28.41 
Save(3%)
Add to cart
Novex™ IEF Sample Buffer pH 3-7 (2X)
Catalog numberLC5371
Price (USD)/ 25 mL
27.65
Online exclusive
28.41 
Save(3%)
-
Add to cart
Novex® IEF Sample Buffer pH 3-7 (2X) is optimized pre-mixed IEF sample buffer for using with Novex® pH 3-7 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSample Loading Buffers
Concentration2 X
Gel TypeIEF
pH Range3 to 7
Product TypeIEF Sample Buffer
Gel CompatibilityNovex™ IEF Gels
Product LineNovex™
Quantity25 mL
Shipping ConditionWet Ice
Unit Size25 mL
Contents & Storage
Store in refrigerator (2–8°C).