Novex™ IEF Cathode Buffer pH 3-7 (10X)
Novex™ IEF Cathode Buffer pH 3-7 (10X)
Invitrogen™

Novex™ IEF Cathode Buffer pH 3-7 (10X)

Novex™ IEF Cathode Buffer pH 3-7 (10X) is optimized pre-mixed IEF cathod buffer for using with Novex™ pH 3-7 IEFRead more
Have Questions?
Catalog number LC5370
Price (USD)/ 125 mL
126.65
Online exclusive
133.00 
Save 6.35 (5%)
-
Add to cart
Price (USD)/ 125 mL
126.65
Online exclusive
133.00 
Save 6.35 (5%)
Add to cart
Novex™ IEF Cathode Buffer pH 3-7 (10X)
Catalog numberLC5370
Price (USD)/ 125 mL
126.65
Online exclusive
133.00 
Save 6.35 (5%)
-
Add to cart
Novex™ IEF Cathode Buffer pH 3-7 (10X) is optimized pre-mixed IEF cathod buffer for using with Novex™ pH 3-7 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.

Usage note: The IEF cathode buffer solutions contain a reagent that varies from white to light yellow in color. This color variation may produce a similar color variation in the final solution, from colorless to yellowish in tone. This color will not affect product performance.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferAnode & Cathode Buffers
Concentration10 X
Gel TypeIEF
pH Range3 to 7
Product TypeIEF Cathode Buffer
Gel CompatibilityNovex™ IEF Gels
Product LineNovex™
Quantity125 mL
Shipping ConditionWet Ice
Unit Size125 mL
Contents & Storage
Store in refrigerator (2–8°C).