Nitrocellulose/Filter Paper Sandwiches, 0.45 μm, 8.6 x 13.5 cm
Invitrogen™
Nitrocellulose/Filter Paper Sandwiches, 0.45 μm, 8.6 x 13.5 cm
Nitrocellulose/Filter Paper Sandwiches, 0.45 μm, are high-quality membranes ideal for blotting proteins and nucleic acids. The 0.45-μm pore size isRead more
Have Questions?
Catalog number LC2006
Price (USD)/ 16 membrane/filter paper sandwiches
274.65
Online exclusive
288.00
Save 13.35 (5%)
-
Add to cart
Price (USD)/ 16 membrane/filter paper sandwiches
274.65
Online exclusive
288.00
Save 13.35 (5%)
Add to cart
Nitrocellulose/Filter Paper Sandwiches, 0.45 μm, 8.6 x 13.5 cm
Catalog numberLC2006
Price (USD)/ 16 membrane/filter paper sandwiches
274.65
Online exclusive
288.00
Save 13.35 (5%)
-
Add to cart
Nitrocellulose/Filter Paper Sandwiches, 0.45 μm, are high-quality membranes ideal for blotting proteins and nucleic acids. The 0.45-μm pore size is ideal for the transfer of most proteins (>20 kDa) and nucleic acids (>300 bp). The included extra-thick absorbent blotting paper (2.5 mm) accommodates blotting set-up using semi-dry transfer apparatuses.
Features: • Membrane: 100% pure nitrocellulose • Filter paper thickness: 2.5 mm • Binding capacity: ∼80 μg/cm2 of proteins • Binding interactions: hydrophobic and electrostatic • Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling • Supplied in a pre-cut, pre-assembled membrane/filter paper sandwich
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Length (Metric)13.5 cm
Width (Metric)8.6 cm
DescriptionNitrocellulose/Filter Paper Sandwich, 0.45μm pore size