LIVE/DEAD™ Yeast Viability Kit
LIVE/DEAD™ Yeast Viability Kit
Invitrogen™

LIVE/DEAD™ Yeast Viability Kit

The LIVE/DEAD® Yeast Viability Kit combines a novel two-color fluorescent probe for yeast viability, FUN® 1, with a fluorescent fungalRead more
Have Questions?
Catalog number L7009
Price (USD)/ 1 kit
698.00
-
Add to cart
Price (USD)/ 1 kit
698.00
Add to cart
LIVE/DEAD™ Yeast Viability Kit
Catalog numberL7009
Price (USD)/ 1 kit
698.00
-
Add to cart
The LIVE/DEAD® Yeast Viability Kit combines a novel two-color fluorescent probe for yeast viability, FUN® 1, with a fluorescent fungal surface labeling reagent Calcofluor® White M2R. Plasma membrane integrity and metabolic function of fungi are required to convert the yellow-green-fluorescent intracellular staining of FUN® 1 into red-orange intravacuolar structures; Calcofluor White M2R labels cell-wall chitin with blue-fluorescence regardless of metabolic state.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeYeast Cells
ColorYellow-green, red-orange, blue
DescriptionLIVE/DEAD™ Yeast Viability Kit
Detection MethodFluorescence
For Use With (Application)Viability Assay
For Use With (Equipment)Fluorescence Microscope, Fluorometer, Flow Cytometer, Microplate Reader
Product TypeYeast Viability Kit
Dye TypeOther Label(s) or Dye(s)
Emission530, 620, 500, 550 nm
Excitation Wavelength Range485, 450 nm
FormatTube(s), 96-well plate, Slide(s)
Product LineLIVE/DEAD™
Quantity1 kit
Shipping ConditionRoom Temperature
Unit Size1 kit
Contents & Storage
Store in freezer at -5°C to -30°C and protect from light.