LIVE/DEAD™ Violet Viability/Vitality Kit, for 405 nm excitation
LIVE/DEAD™ Violet Viability/Vitality Kit, for 405 nm excitation
Invitrogen™

LIVE/DEAD™ Violet Viability/Vitality Kit, for 405 nm excitation

The LIVE/DEAD™ Violet Viability/Vitality Kit provides a two-color fluorescence cell viability and vitality assay that is based on the simultaneousRead more
Have Questions?
Catalog number L34958
Price (USD)/ 1 kit
640.00
-
Add to cart
Price (USD)/ 1 kit
640.00
Add to cart
LIVE/DEAD™ Violet Viability/Vitality Kit, for 405 nm excitation
Catalog numberL34958
Price (USD)/ 1 kit
640.00
-
Add to cart
The LIVE/DEAD™ Violet Viability/Vitality Kit provides a two-color fluorescence cell viability and vitality assay that is based on the simultaneous determination of live and dead cells with two probes that measure recognized parameters of cell health: plasma membrane integrity and intracellular esterase activity. Calcein violet AM and aqua-fluorescent reactive dye are optimal dyes for this application; both dyes utilize the violet laser allowing other laser lines to be used for conventional fluorochromes.

View a selection guide for all Nonfixable Viability Dyes for Flow Cytometry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityImpermeant
Cell TypeEukaryotic Cells
ColorAqua, Violet
DescriptionLIVE/DEAD™ Violet Viability/Vitality Kit, for 405 nm excitation
Detection MethodFluorescence
For Use With (Application)Viability Assay
For Use With (Equipment)Flow Cytometer
Product TypeStain
Dye TypeOther Label(s) or Dye(s)
EmissionAqua-fluorescent reactive dye: 367/526 nm
Calcein violet stain: 400/452 nm
Excitation Wavelength Range367/400 nm
FormLyophilized
FormatTube(s)
Product LineLIVE/DEAD™
Quantity1 kit
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
Unit Size1 kit
Contents & Storage
Contains 5 vials of calcein violet stain (25 μg/vial), 5 vials of aqua-fluorescent reactive dye, and 1 vial of DMSO.

Store in freezer (-5°C to -30°C) and protect from light.