Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 649
Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 649
Invitrogen™

Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 649

Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 649, is a bright deep-red/far-red LEL-fluorophore conjugate. The tomato lectin LEL is widelyRead more
Have Questions?
Catalog number L32472
Price (USD)/ 1 mg
197.65
Online exclusive
201.00 
Save 3.35 (2%)
-
Add to cart
Quantity:
1 mg
Price (USD)/ 1 mg
197.65
Online exclusive
201.00 
Save 3.35 (2%)
Add to cart
Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 649
Catalog numberL32472
Price (USD)/ 1 mg
197.65
Online exclusive
201.00 
Save 3.35 (2%)
-
Add to cart
Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 649, is a bright deep-red/far-red LEL-fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research. LEL-DyLight 649 can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.

View complete list of fluorescent dye-conjugated lectins ›

• Very bright DyLight 649 conjugate detected with traditional Cy5/deep-red filter set, Ex/Em: 655/670 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
• Inhibiting/eluting sugar: chitin hydrolysate
• Blood group specificity: non-specific
• Unconjugated lectin molecular weight: 71 KDa.
• Immunofluorescence working concentration: 5–20 μg/mL

LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 mg
SourceTomato
ConjugateDyLight 649
Label TypeDyLight Fluors
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
FormSolution
Protein SubtypeLectins
Product LineDyLight™
Unit Size1 mg
Contents & Storage
When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.