LIVE/DEAD™ Viability/Cytotoxicity Assay Kit (Green/Deep Red)
LIVE/DEAD™ Viability/Cytotoxicity Assay Kit (Green/Deep Red)
Invitrogen™

LIVE/DEAD™ Viability/Cytotoxicity Assay Kit (Green/Deep Red)

The LIVE/DEAD Viability/Cytotoxicity Assay Kit (Green/Deep Red) is a two-color fluorescent cell viability assay that simultaneously identifies live and deadRead more
Have Questions?
Catalog number L32250
Price (USD)/ 1 kit
331.65
Online exclusive
336.00 
Save 4.35 (1%)
-
Add to cart
Price (USD)/ 1 kit
331.65
Online exclusive
336.00 
Save 4.35 (1%)
Add to cart
LIVE/DEAD™ Viability/Cytotoxicity Assay Kit (Green/Deep Red)
Catalog numberL32250
Price (USD)/ 1 kit
331.65
Online exclusive
336.00 
Save 4.35 (1%)
-
Add to cart
The LIVE/DEAD Viability/Cytotoxicity Assay Kit (Green/Deep Red) is a two-color fluorescent cell viability assay that simultaneously identifies live and dead cells. Live cells are labeled with calcein dye and emit a green fluorescence, while dead cells are labeled with SYTOX Deep Red dye and emit a red fluorescence. These dyes can be multiplexed with other orange/red fluorophores with compatible filter sets.

• Determines cellular viability with two-color fluorescence
• Detect live cells with GFP/FITC filter set and dead cells with Deep Red/CY5 filter set
• Multiplexable with orange/red fluorophores such as RFP, mCherry, Mitotracker Red/Orange, Lysotracker Red, TMRM, and others

Live cells are distinguished by the presence of ubiquitous intracellular esterase activity, determined by the enzymatic conversion of the virtually non-fluorescent cell-permeant Calcein AM to the intensely fluorescent calcein. Calcein is well retained within live cells, producing an intense uniform green fluorescence in live cells (ex/em ∼495/∼515 nm). Calcein can be detected with a traditional FITC/GFP filter set. SYTOX Deep Red enters cells with damaged membranes and increases its fluorescence many-fold upon binding to DNA. Dead or plasma membrane-compromised nuclei fluorescence brightly and can be detected with a CY5/Deep Red filter set (ex/em maxima: 660/682 nm).

The kit is suitable for use with fluorescence microscopes or high content analysis (HCA) fluorescence multiwall plate scanners and easily adaptable for use other fluorescence detection systems. The assay principles are general and applicable to most eukaryotic cell types, including adherent and non-adherent cells.

Specifications
ColorGreen, Deep Red
DescriptionLIVE/DEAD Viability/Cytotoxicity Assay Kit (Green/Deep Red)
Detection MethodFluorescence
For Use With (Application)Viability Assay
For Use With (Equipment)Fluorescence Microscope, High Content Analysis Instrument, Other Fluorescence Detection Systems
Product TypeM25
Emission515, 682 nm
Excitation Wavelength Range495, 660 nm
FormatKit
IndicatorCalcein, AM (live cell indicator), SYTOX™ Deep Red Nucleic Acid Stain (dead cell indicator)
Product LineLIVE/DEAD
Quantity2 x 40 μL Vials
Shipping ConditionRoom Temperature
TargetViability
Unit Size1 kit
Contents & Storage
• 1 vial of Calcein AM (live cell indicator), 40 μL, provided as a 2000X (4 mM) solution in DMSO
• 1 vial of SYTOX Deep Red Nucleic Acid Stain (dead cell indicator), 40 μL, provided as a 2000X (500 μM) solution in DMSO

Store at ≤-20°C, upright in dry place. Protect from light.