GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 Cells

GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 cells contain the human 5-hydroxytrptamine (serotonin) receptor 2C (HTR2C), (Accession No. NM_000868.1) stably integrated into the CellSensor™Read more
Have Questions?
Catalog number K1877
Price (USD)/ 8 x 106 cells⁄vial
10,500.00
-
Add to cart
Price (USD)/ 8 x 106 cells⁄vial
10,500.00
Add to cart
GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 Cells
Catalog numberK1877
Price (USD)/ 8 x 106 cells⁄vial
10,500.00
-
Add to cart

GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 cells contain the human 5-hydroxytrptamine (serotonin) receptor 2C (HTR2C), (Accession No. NM_000868.1) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the NFAT Response Element. The GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of 5-Hydroxytrptamine. In addition, GeneBLAzer™ HTR2C-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_000868.1
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
For Use With (Application)FRET
Human Protein ClassG-Protein Coupled Receptor (GPCR)
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity8 x 106 Cells
ReadoutEnd Point
Reporter GeneBLA (Beta-Lactamase)
Target EntryHTR2C
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size8 x 106 cells⁄vial
Contents & Storage
GeneBLAzer™ cell lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.