Tango™ GPR35-bla U2OS cells contain the human G-protein coupled receptor 35 (GPR35) linked to a TEV protease site and aRead more
Have Questions?
Catalog number K1872
Price (USD)/ 1 vial
10,530.00
-
Add to cart
Price (USD)/ 1 vial
10,530.00
Add to cart
Tango™ GPR35-bla U2OS Cells
Catalog numberK1872
Price (USD)/ 1 vial
10,530.00
-
Add to cart
Tango™ GPR35-bla U2OS cells contain the human G-protein coupled receptor 35 (GPR35) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin⁄TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ GPR35-bla U2OS cells have been functionally validated for Z’ factor and EC50 concentrations of Zaprinast. In addition, Tango™ GPR35-bla U2OS cells have been tested for assay performance under variable conditions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_005301.2
Assay EntryCell-based beta-arrestin recruitment
Cell StateDividing Cells
Cytoplasmic TailNative Tail
Detection MethodFluorescence
For Use With (Application)FRET
Human Protein ClassG-Protein Coupled Receptor (GPCR)
Population StatusSingle Cell Clone Origin
Product LineTango™
Quantity1 vial
ReadoutEnd Point
Target EntryGPR35
Cell LineU2OS
System TypeTango™
Druggable TargetGPCR
Unit Size1 vial
Contents & Storage
Tango™ GPR35-bla U2OS cells cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.