GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells contain the human Purinergic Receptor P2Y4 (P2RY4), stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the NFAT Response Element. The GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of UTP. In addition, GeneBLAzer™ P2RY4-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.