Tango™ MLNR-bla U2OS Cells
Product Image
Thermo Scientific™

Tango™ MLNR-bla U2OS Cells

Tango™ MLNR-bla U2OS cells contain the human Motilin Receptor (MLNR) linked to a TEV protease site and a Gal4-VP16 transcriptionRead more
Have Questions?
Catalog number K1862
Price (USD)/ 1 vial
10,580.00
-
Add to cart
Price (USD)/ 1 vial
10,580.00
Add to cart
Tango™ MLNR-bla U2OS Cells
Catalog numberK1862
Price (USD)/ 1 vial
10,580.00
-
Add to cart

Tango™ MLNR-bla U2OS cells contain the human Motilin Receptor (MLNR) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ MLNR-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of Motilin.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_001507
Assay EntryCell-based beta-arrestin recruitment
Cell StateDividing Cells
Cytoplasmic TailTail from AVPR2 GPCR
Detection MethodFluorescence
For Use With (Application)FRET
Human Protein ClassG-Protein Coupled Receptor (GPCR)
Population StatusSingle Cell Clone Origin
Product LineTango™
Quantity1 vial
ReadoutEnd Point
Target EntryMLNR
Cell LineU2OS
System TypeTango™
Druggable TargetGPCR
Unit Size1 vial
Contents & Storage
Tango™ MLNR-bla U2OS cells are shipped on dry ice, 8 x 106 cells/vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.