GeneBLAzer™ PROKR2-Gqi5-NFAT-bla CHO-K1 cells contain the human prokineticin receptor 2 (PROKR2), (Accession No. NM_144773) stably integrated into the GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqi5-NFAT-bla CHO-K1 cells (Cat. No. K1725) contain a beta-lactamase reporter gene under control of the NFAT Response Element. The GeneBLAzer™ PROKR2-Gqi5-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Prokineticin (EG-VGEF)