GeneBLAzer® GCGR-CRE-bla CHO-K1 cells contain the human glucagon receptor (GCGR), (Accession # NM_000160) stably integrated into the CellSensor® CRE-bla CHO-K1Read more
Have Questions?
Catalog number K1855
Price (USD)/ 1 vial
10,500.00
-
Add to cart
Price (USD)/ 1 vial
10,500.00
Add to cart
GeneBLAzer™ GCGR-CRE-bla CHO-K1 Cells
Catalog numberK1855
Price (USD)/ 1 vial
10,500.00
-
Add to cart
GeneBLAzer® GCGR-CRE-bla CHO-K1 cells contain the human glucagon receptor (GCGR), (Accession # NM_000160) stably integrated into the CellSensor® CRE-bla CHO-K1 cell line. CellSensor® CRE-bla CHO-K1 (Cat No. K1535) cells contain a beta-lactamase reporter gene under control of the CRE Response Element. The GCGR-CRE-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of Glucagon. In addition, GCGR-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions.
For Research Use Only. Not for use in diagnostic procedures.
Human Protein ClassG-Protein Coupled Receptor (GPCR)
Population StatusPool of Stable Cells
Product LineGeneBLAzer™
Quantity1 vial
ReadoutEnd Point
Target EntryGCGR
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size1 vial
Contents & Storage
GeneBLAzer™ GCGR-CRE-bla CHO-K1 cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.