Tango™ GPR1-bla U2OS Cells
Product Image
Thermo Scientific™

Tango™ GPR1-bla U2OS Cells

Tango™ GPR1-bla U2OS cells contain the human G protein-coupled receptor 1 (GPR1) linked to a TEV protease site and aRead more
Have Questions?
Catalog number K1839
Price (USD)/ 8 x 106 cells
10,580.00
-
Add to cart
Price (USD)/ 8 x 106 cells
10,580.00
Add to cart
Tango™ GPR1-bla U2OS Cells
Catalog numberK1839
Price (USD)/ 8 x 106 cells
10,580.00
-
Add to cart

Tango™ GPR1-bla U2OS cells contain the human G protein-coupled receptor 1 (GPR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ GPR1-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of Chemerin (Figure 1). In addition, Tango™ GPR1-bla U2OS cells have been tested for assay performance under variable conditions.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_005279.3
Assay EntryCell-based beta-arrestin recruitment
Cell StateDividing Cells
Cytoplasmic TailNative Tail
Detection MethodFluorescence
For Use With (Application)FRET
Gene SymbolGPR1
Human Protein ClassG-Protein Coupled Receptor (GPCR)
Population StatusSingle Cell Clone Origin
Product LineTango™
Quantity8 x 106 Cells
ReadoutEnd Point
Target EntryGPR1
Cell LineU2OS
System TypeTango™
Druggable TargetGPCR
Unit Size8 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• Tango™ GPR1-UAS-bla CHO-K1 cells
• Tango™ GPR1-UAS-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The Tango™ GPR1-UAS-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.