GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells
Product Image
Thermo Scientific™

GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells

GeneBLAzer™ ADORA3-Ga15-NFAT-bla CHO-K1 cells contain the human Adenosine A3 Receptor (ADORA3) stably integrated into the GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cell line.Read more
Have Questions?
Catalog number K1837
Price (USD)/ 8 x 106 cells
10,580.00
-
Add to cart
Price (USD)/ 8 x 106 cells
10,580.00
Add to cart
GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells
Catalog numberK1837
Price (USD)/ 8 x 106 cells
10,580.00
-
Add to cart

GeneBLAzer™ ADORA3-Ga15-NFAT-bla CHO-K1 cells contain the human Adenosine A3 Receptor (ADORA3) stably integrated into the GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cell line. The GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cells (Cat. No. K1537) contain a beta-lactamase reporter gene under control of the NFAT Response Element and a promiscuous G protein, Ga15. GeneBLAzer™ ADORA3-Ga15-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of NECA.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_000677.2
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
For Use With (Application)FRET
Gene SymbolADORA3
Human Protein ClassG-Protein Coupled Receptor (GPCR)
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity8 x 106 Cells
ReadoutEnd Point
Target EntryADORA3
Cell LineCHO-K1
System TypeTango™
Druggable TargetGPCR
Unit Size8 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells
• GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.