The GeneBLAzer™ D5-CRE-bla CHO-K1 cells contain the human Dopamine Receptor 5 (D5) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase reporter gene under control of the CRE response element.
The GeneBLAzer™ D5-CRE-bla CHO-K1 cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, GeneBLAzer™ D5-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.