GeneBLAzer™ GLP1R-CRE-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ GLP1R-CRE-bla CHO-K1 Cells

The GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells contain the human Glucagon-like Peptide 1 Receptor (GLP1R) stably integrated into the CellSensor™ CRE-bla CHO-K1Read more
Have Questions?
Catalog number K1783
Price (USD)/ 8 x 106 cells
10,380.00
-
Add to cart
Price (USD)/ 8 x 106 cells
10,380.00
Add to cart
GeneBLAzer™ GLP1R-CRE-bla CHO-K1 Cells
Catalog numberK1783
Price (USD)/ 8 x 106 cells
10,380.00
-
Add to cart

The GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells contain the human Glucagon-like Peptide 1 Receptor (GLP1R) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase reporter gene under control of the CRE response element.

The GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_002062.2
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
Gene SymbolGLP1R
Population StatusPool of Stable Cells
Product LineGeneBLAzer™
Protein FamilyGPCR (G-Protein Coupled Receptors)
Quantity8 x 106 Cells
ReadoutEnd Point
Reporter GeneBLA (Beta-Lactamase)
Target EntryGLP1R
TypeCHO-K1 Cells
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size8 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells
• GeneBLAzer™ GLP1R-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ GLP1R-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.