GeneBLAzer™ T-REx™ G2A-NFAT-bla Freestyle HEK 293F cells contain the human GPR132 (G2A), (Accession No. NP_037477) stably integrated into the CellSensor™ NFAT-bla Freestyle HEK 293F cell line. CellSensor™ NFAT-bla Freestyle HEK 293F cells (Cat. no. K1725) contain a beta-lactamase reporter gene under control of the NFAT Response Element.
The GeneBLAzer™ T-REx™ G2A-NFAT-bla Freestyle HEK 293F cells are functionally validated for Z' and EC50 concentrations of dDAVP. In addition, GeneBLAzer™ GeneBLAzer™ T-REx™ G2A-NFAT-bla Freestyle HEK 293F cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
G2A is an orphan GPCR. Upon overexpession G2A is constitutively active. This is an assay for the orphan GPCR based upon constitutive activity. To use the assay to detect inverse agonists, high amounts of doxycycline should be added. To use the assay to detect agonists, low amounts of doxycycline should be added.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.