GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 Cells

GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 cells contain the human Complement Component 5a Receptor 1 (C5aR1) (Accession No. NM_000710) stably integrated into theRead more
Have Questions?
Catalog number K1746
Price (USD)/ 2 x 106 cells
10,500.00
-
Add to cart
Price (USD)/ 2 x 106 cells
10,500.00
Add to cart
GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 Cells
Catalog numberK1746
Price (USD)/ 2 x 106 cells
10,500.00
-
Add to cart

GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 cells contain the human Complement Component 5a Receptor 1 (C5aR1) (Accession No. NM_000710) stably integrated into the GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gα15-NFAT-bla CHO-K1 (Cat. No. K1537) contains a beta-lactamase reporter gene under control of a NFAT response element and a promiscuous G Protein, Gα15, stably integrated into CHO-K1 cells.

The GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of C5a. In addition, GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_001736.2
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
For Use With (Application)FRET
Gene SymbolC5aR1
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity2 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryC5AR1
TypeCHO-K1 Cells
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size2 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 cells
• GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ C5aR1-Gα15-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.