GeneBLAzer™ MC5R-CRE-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ MC5R-CRE-bla CHO-K1 Cells

GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells contain the human Melanocortin Receptor 5 (MC5R) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.Read more
Have Questions?
Catalog number K1740
Price (USD)/ 2 x 106 cells
10,480.00
-
Add to cart
Price (USD)/ 2 x 106 cells
10,480.00
Add to cart
GeneBLAzer™ MC5R-CRE-bla CHO-K1 Cells
Catalog numberK1740
Price (USD)/ 2 x 106 cells
10,480.00
-
Add to cart

GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells contain the human Melanocortin Receptor 5 (MC5R) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.

The GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of NPD-α MSH In addition, GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

MC5R is expressed in skin, muscle, lung, liver, spleen, and adrenal tissue. MC5R is Gs-coupled and binds all of the melanocortin peptides and ACTH. A recent study suggests that MC5R is involved in fat storage in the sebocyte cells of the skin. Another study in mice links MC5R to aggressive behavior. Mice deficient in MC5R acted less aggressively when treated with alpha-MSH.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_005913.1
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
For Use With (Application)FRET
Gene SymbolMC5R
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity2 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryMC5R
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size2 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells
• GeneBLAzer™ MC5R-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.