GeneBLAzer™ M1-NFAT-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ M1-NFAT-bla CHO-K1 Cells

GeneBLAzer™ M1-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla CHO-K1Read more
Have Questions?
Catalog number K1735
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart
Price (USD)/ 2 x 106 cells
10,580.00
Add to cart
GeneBLAzer™ M1-NFAT-bla CHO-K1 Cells
Catalog numberK1735
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart

GeneBLAzer™ M1-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase (bla) reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.

M1-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of carbachol. In addition, M1-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

In the brain, M1 activation mediates 'slow' neuronal excitability. Cortical and hippocampal muscarinic receptors are thought to be important in the attentional aspects of cognition. The predominant receptor subtypes in these brain areas are M1, M3, and M4. Therefore, M1 is a potential target for cognition, Alzheimer's, dementia, and schizophrenia. Studies on knock-out mouse models of M1 are also beginning to reveal potential functions of the receptor. Additional information on the muscarinic receptors can be found in reviews.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_000738
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
For Use With (Application)FRET
Gene SymbolM1
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity2 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryM1
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size2 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ M1-NFAT-bla CHO-K1 cells
• GeneBLAzer™ M1-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ M1-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.