For academic pricing please login or contact us at discoverysciences@invitrogen.com.
GeneBLAzer™ PTGER2-NFAT-bla CHO-K1 cells contain the human Prostaglandin E Receptor 2 (PTGER2), (Accession No. NM_000956.2) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of the NFAT Response Element.
GeneBLAzer™ PTGER2-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.