GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells contain the human KiSS1 receptor (GPR54) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
The GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Metastin. In addition, GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
The GPR54 receptor has been shown to be involved in tumor progression and metastasis, and in the development and functional regulation of the gonads.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.