GeneBLAzer™ HTR7-CRE-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ HTR7-CRE-bla CHO-K1 Cells

GeneBLAzer™ HTR7 CHO-K1 DA (Division Arrested) cells and HTR7-CRE-bla CHO-K1 cells contain the human 5-hydroxytryptamine (serotonin) receptor 7 (Accession No.Read more
Have Questions?
Catalog number K1704
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart
Price (USD)/ 2 x 106 cells
10,580.00
Add to cart
GeneBLAzer™ HTR7-CRE-bla CHO-K1 Cells
Catalog numberK1704
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart

GeneBLAzer™ HTR7 CHO-K1 DA (Division Arrested) cells and HTR7-CRE-bla CHO-K1 cells contain the human 5-hydroxytryptamine (serotonin) receptor 7 (Accession No. NM_019860.2) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1535) contain a beta-lactamase (bla) reporter gene under control of the Cyclic AMP Response Element (CRE).

GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of 5-hydroxytryptamine (5-HT). In addition, GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_019860.2
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
For Use With (Application)FRET
Gene SymbolHTR7
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity2 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryHTR7
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size2 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells
• GeneBLAzer™ HTR7-CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ HTR7-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.