The CellSensor™ ISRE-bla RA-1 Cell Line contains the beta-lactamase (bla) gene under the control of the interferon-stimulated response element (ISRE). This cell line was engineered by lentiviral transduction of an ISRE-bla construct into RA-1 cells. Flow cytometry was then used to isolate a pool of cells responsive to polyinosinic-polycytidylic acid (poly I:C) stimulation. ISRE-bla RA-1 cells have been tested for assay performance using variable assay conditions, including DMSO concentration, cell number, stimulation time, substrate loading time, as well as validated for Z'-factor and EC50 concentrations of poly I:C. Additional testing data using alternate stimuli are also available. Academic and non-profit customers, please inquire for special pricing.