Interferon cell signaling is a critical pathway involved in viral defense and autoimmune disease. The CellSensor™ ISRE-bla HEK 293T cell line is responsive to interferon alpha and can be used to probe the JAK/STAT signaling pathway. The CellSensor™ ISRE-bla HEK 293T cell line contains the β-lactamase reporter gene under the control of the interferon stimulated response element (ISRE). This cell line was engineered by transduction of the ISRE-bla construct into HEK 293T cells by lentivirus. Flow cytometry was then used to isolate a pool of clones responsive to interferon alpha a stimulation. A dose response curve for this assay against interferon alpha is available in the validation packet. Academic and non-profit customers, please inquire for special pricing.