CellSensor™ irf1-bla Ba/F3 Cell Line
Product Image
Thermo Scientific™

CellSensor™ irf1-bla Ba/F3 Cell Line

Jak/Stat signaling pathways play essential roles in the cellular responses to distinct cytokines. One of the Jak/Stat pathways, Jak2/Stat5, mediatesRead more
Have Questions?
Catalog number K1654
Price (USD)/ 1 vial
10,530.00
-
Add to cart
Price (USD)/ 1 vial
10,530.00
Add to cart
CellSensor™ irf1-bla Ba/F3 Cell Line
Catalog numberK1654
Price (USD)/ 1 vial
10,530.00
-
Add to cart

Jak/Stat signaling pathways play essential roles in the cellular responses to distinct cytokines. One of the Jak/Stat pathways, Jak2/Stat5, mediates cell proliferation in response to Interleukin-3 (IL-3), prolactin, erythropoietin (Epo), and granulocyte - macrophage colony stimulating factor (GM-CSF). JAK2 gene knockout causes embryonic lethality due to defective erythropoiesis, suggesting that the Jak2/Stat5 pathway plays an important role in red blood cell formation. The recent discovery of an activating mutation in JAK2 (V617F) present in a high percentage of myeloproliferative disease (MPD) patients suggests that the Jak2/Stat5 pathway may be a potential therapeutic target for certain forms of MPD. The activated Stat5 transcription factor recognizes and binds to a specific palindromic DNA sequence found in the promoter region of β-casein, interferon regulatory factor-1 (irf-1), and a number of other genes.

The CellSensor™ irf1-bla BA/F3 Cell Line contains a beta-lactamase reporter gene under control of the irf-1 response element stably integrated into BA/F3 cells. This cell line is validated for EC50 and Z' - factor under optimized conditions using mouse IL-3 (mIL-3). This cell line has also been tested under variable experimental conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time. Responsiveness to Jak Inhibitor 1, a small molecule inhibitor, was also tested. CellSensor™ irf1-bla BA/F3 cells were treated with mIL-3 in triplicate over the indicated concentration range in a 384-well format. Cells were incubated for 5 hr. with mIL-3 agonist in 0.5% DMSO and then combined with LiveBLAzer™-FRET B/G Substrate (CCF4-AM) for 2.5 hr. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. The Response Ratios for each replicate were plotted against the indicated concentrations of mIL-3. CellSensor™ irf1-bla BA/F3 cells were treated with Jak Inhibitor 1 for 30 min over the indicated concentration range in a 384 - well format. Cells were then incubated with mIL - 3 agonist in 0.5% DMSO for 5 hr. and then combined with LiveBLAzer™ - FRET B/G Substrate (CCF4-AM) for 2.5 hr. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. The emission ratios were plotted against the indicated concentrations of Jak Inhibitor 1. Academic and non-profit customers, please inquire for special pricing.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Product LineCellSensor™
Quantity1 Vial
Target EntrySTAT5, JAK/STAT (IL-3) pathway, JAK2
System TypeCellSensor™
Druggable TargetSignaling Pathway
Unit Size1 vial
Contents & Storage
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.