CellSensor™ IL1-bla ECV304 Cell Line
Product Image
Thermo Scientific™

CellSensor™ IL1-bla ECV304 Cell Line

The CellSensor™ IL-1-bla ECV304 cell line is an adherent human urinary bladder cell line used to measure the (IL-1) signalingRead more
Have Questions?
Catalog number K1645
Price (USD)/ 1 vial
10,580.00
-
Add to cart
Price (USD)/ 1 vial
10,580.00
Add to cart
CellSensor™ IL1-bla ECV304 Cell Line
Catalog numberK1645
Price (USD)/ 1 vial
10,580.00
-
Add to cart

The CellSensor™ IL-1-bla ECV304 cell line is an adherent human urinary bladder cell line used to measure the (IL-1) signaling pathway via transcriptional regulation of the beta-lactamase gene. IL-1 cytokine cell signaling is a critical pathway involved in immune and inflammatory responses. This cell line was constructed using GenomeScreen™ technology utilizing a promoterless beta-lactamase reporter gene randomly integrated into the genome of the IL-1-bla ECV304 cell line. Sequential rounds of sorting were performed to isolate cell clones containing tagged genes induced by IL-1. A dose response assay was run with this cell line using IL-1 as the agonist. Academic and non-profit customers, please inquire for special pricing.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Product LineCellSensor™
Quantity1 Vial
Target EntryInterleukin 1 pathway, IRAK
System TypeCellSensor™
Druggable TargetSignaling Pathway
Unit Size1 vial
Contents & Storage
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.