CellSensor™ CRE-bla FreeStyle™ 293F Cell Line
Product Image
Thermo Scientific™

CellSensor™ CRE-bla FreeStyle™ 293F Cell Line

The CellSensor™ CRE-bla FreeStyle™ 293F cell line contains a beta-lactamase reporter gene under control of the CRE response element stablyRead more
Have Questions?
Catalog number K1636
Price (USD)/ 1 vial
10,580.00
-
Add to cart
Price (USD)/ 1 vial
10,580.00
Add to cart
CellSensor™ CRE-bla FreeStyle™ 293F Cell Line
Catalog numberK1636
Price (USD)/ 1 vial
10,580.00
-
Add to cart

The CellSensor™ CRE-bla FreeStyle™ 293F cell line contains a beta-lactamase reporter gene under control of the CRE response element stably integrated into FreeStyle™ 293F cells. To construct this cell line, the CRE -bla construct was transduced into FreeStyle™ 293F cells by lentivirus. Flow cytometry was used to isolate cells responsive to forskolin. Assay development experiments performed on this cell line included DMSO tolerance, cell number, stimulation time and substrate loading time. Functional validation of this cell line under optimized conditions included and Z' and EC50 determinations using forskolin as a primary agonist. The CellSensor™ CRE-bla FreeStyle™ 293F cell line is responsive to forskolin and can be used to build specific G-protein coupled receptor (GPCR) assays after transfection of additional genes of interest or to detect changes in cellular cAMP levels. This cell line can be adapted for high-throughput screening for agonists or antagonists with compound libraries. Candidate drugs can be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Product LineCellSensor™
Quantity1 Vial
Target EntryPKA, cAMP, cAMP/PKA pathway
System TypeCellSensor™
Druggable TargetSignaling Pathway
Unit Size1 vial
Contents & Storage
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.