CellSensor™ ARE-bla Hep G2 Cell Line
Product Image
Invitrogen™

CellSensor™ ARE-bla Hep G2 Cell Line

The CellSensor® ARE-bla Hep G2 cell line is responsive to tBHQ and Sulforaphane and can be used for analyzing theRead more
Have Questions?
Catalog number K1633
Price (USD)/ 1 vial
14,810.00
-
Add to cart
Price (USD)/ 1 vial
14,810.00
Add to cart
CellSensor™ ARE-bla Hep G2 Cell Line
Catalog numberK1633
Price (USD)/ 1 vial
14,810.00
-
Add to cart
The CellSensor® ARE-bla Hep G2 cell line is responsive to tBHQ and Sulforaphane and can be used for analyzing the Nrf2/antioxidant response signaling pathway using the GeneBLAzer® Loading Substrates. Reactive oxygen species (ROS) can damage biological macromolecules and are detrimental to cellular health. Electrophilic compounds, xenobiotics and antioxidants are sources of reactive oxygen species, creating oxidative stress that can harm cells. Enzymes are involved in the Phase II detoxification of xenobiotics to reduce cellular stress include glutathione transferases, quinone reductase, epoxide hydrolase, heme oxygenase, UDP-glucuronosyl transferases, and gamma-glutamylcysteine synthetase. Expression of these genes protects cells from oxidative damage and can prevent mutagenesis and cancer. Transcription of these enzymes is coordinately regulated through antioxidant response elements (AREs). Nrf2 (NF-E2-related factor 2) and Nrf1 are transcription factors that bind to AREs and activate these genes. The CellSensor™ ARE-bla Hep G2 cell line contains a beta-lactamase reporter gene under control of the Antioxidant Response Element (ARE) stably integrated into Hep G2 cells. To construct this cell line, the ARE-bla construct was transduced into Hep G2 cells. Flow cytometry was used to isolate cells responsive to tert-butylhydroquinone (tBHQ). This cell line is tested under various experimental conditions, including DMSO concentration, cell number, stimulation time, substrate loading time, and functionally validated for Z'; and EC50 concentrations of tBHQ. Academic and non-profit customers, please inquire for special pricing.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Product LineCellSensor™
Quantity1 vial
Target EntryAnti-oxidant response pathway, NRF2
TypeCell Line
Cell LineHep G2
System TypeCellSensor™
Druggable TargetSignaling Pathway
Unit Size1 vial
Contents & Storage
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.