RevertAid H Minus First Strand cDNA Synthesis Kit
For superior performance upgrade to the Maxima H Minus First Strand cDNA Synthesis Kit
Inquire about OEM or Commercial Supply version of this product here.
RevertAid H Minus First Strand cDNA Synthesis Kit
Thermo Scientific™

RevertAid H Minus First Strand cDNA Synthesis Kit

Thermo Scientific RevertAid H Minus First Strand cDNA Synthesis Kit is a complete system for efficient synthesis of first strandRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K1632100 Reactions
K163120 Reactions
Catalog number K1632
Price (USD)/ 100 reactions
489.65
Online exclusive
600.00 
Save 110.35 (18%)
-
Add to cart
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (USD)/ 100 reactions
489.65
Online exclusive
600.00 
Save 110.35 (18%)
Add to cart
RevertAid H Minus First Strand cDNA Synthesis Kit
Catalog numberK1632
Price (USD)/ 100 reactions
489.65
Online exclusive
600.00 
Save 110.35 (18%)
-
Add to cart

Thermo Scientific RevertAid H Minus First Strand cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA from RNA templates. The kit includes RevertAid H Minus Reverse Transcriptase, which has a point mutation that completely eliminates RNase H activity. Therefore, it does not degrade RNA in RNA-DNA hybrids during synthesis of the first strand cDNA.

Features of the RevertAid H Minus First Strand cDNA Synthesis Kit include:
• High yields of full-length first-strand cDNA up to 13 kb
• Increased reaction temperatures of 42–55°C
• Supplied with the recombinant RiboLock RNase Inhibitor
• Complete kit includes oligo(dT)18 and random hexamer primers

Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Construction of full length cDNA libraries
• Antisense RNA synthesis

Additional Features
The recombinant RiboLock RNase Inhibitor, supplied with the kit, effectively protects RNA template from degradation. It is fully compatible with the reverse transcription reaction, as it maintains activity at temperatures up to 55°C. The kit is supplied with both oligo(dT)18 and random hexamer primers. The oligo(dT)18 primer anneals selectively to the poly(A) tail of mRNA. Random hexamer primers do not require the presence of poly(A), therefore, they can be used for transcription of the 5’-end regions of mRNA or for cDNA synthesis using RNA without the poly(A) tail, e.g., microRNAs. Gene-specific primers may also be used with the kit. The first strand cDNA can be directly used as a template in PCR, real-time PCR, or in second strand cDNA synthesis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit
Reaction Speed60 min.
TechniqueReverse Transcription
Optimal Reaction Temperature42°C to 45°C
Reverse TranscriptaseRevertAid H Minus
Ribonuclease H ActivityReduced
Shipping ConditionDry Ice
For Use With (Application)Real Time PCR (qPCR), RT-PCR
Final Product TypeFirst-Strand cDNA
No. of Reactions100 Reactions
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Size (Final Product)Up to 13 kb
Starting MaterialRNA
Unit Size100 reactions
Contents & Storage

• RevertAid H Minus Reverse Transcriptase
• RiboLock RNase Inhibitor
• 5X Reaction Buffer
• dNTP Mix
• Oligo(dT)18 Primer
• Random Hexamer Primer
• Control RNA
• Control Primers
• Nuclease-free water

Store at –20°C.