GeneBLAzer™ RAR-gamma DA Assay Kit
Product Image
Thermo Scientific™

GeneBLAzer™ RAR-gamma DA Assay Kit

GeneBLAzer™RAR gamma DA (Division Arrested) cells and RAR gamma-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the humanRead more
Have Questions?
Catalog number K1559
Price (USD)/ 1 plate
1,522.00
-
Add to cart
Price (USD)/ 1 plate
1,522.00
Add to cart
GeneBLAzer™ RAR-gamma DA Assay Kit
Catalog numberK1559
Price (USD)/ 1 plate
1,522.00
-
Add to cart

GeneBLAzer™RAR gamma DA (Division Arrested) cells and RAR gamma-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoic acid receptor gamma (RAR gamma) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-RAR gamma (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both RAR gamma DA cells and RAR gamma-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of all-trans retinoic acid (ATRA). In addition, RAR gamma-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, and stimulation time.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Alternate Agonists4-hydroxytamoxifen, Chlordane, Toxaphene
Assay EntryCell-based beta lactamase reporter gene
Cell StateDivision-Arrested Cells
Detection MethodFluorescence
Gene SymbolRARγ
Primary AgonistXCT790
Quantity1 Plate
Reporter GeneBLA (Beta-Lactamase)
Target EntryRARG, RAR gamma, NR1B3
TypeDivision-Arrested Cells Assay Kit
Cell Line293T (HEK)
System TypeGeneBLAzer™
Druggable TargetNuclear Receptors
Unit Size1 plate
Contents & Storage
Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate.
• RAR gamma HEK 293T DA cells (K1559A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)