GeneBLAzer™ OPRK1-Gqo5-NFAT-bla CHO-K1 Cells
Product Image
Thermo Scientific™

GeneBLAzer™ OPRK1-Gqo5-NFAT-bla CHO-K1 Cells

The GeneBLAzer™ OPRK-1-Gqo5-NFAT-bla CHO-K1 cells contain the human Opioid Receptor Kappa 1 (OPRK1) (Accession No. NM_000710) stably integrated into theRead more
Have Questions?
Catalog number K1533
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart
Price (USD)/ 2 x 106 cells
10,580.00
Add to cart
GeneBLAzer™ OPRK1-Gqo5-NFAT-bla CHO-K1 Cells
Catalog numberK1533
Price (USD)/ 2 x 106 cells
10,580.00
-
Add to cart

The GeneBLAzer™ OPRK-1-Gqo5-NFAT-bla CHO-K1 cells contain the human Opioid Receptor Kappa 1 (OPRK1) (Accession No. NM_000710) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. The GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells (Cat. No. K1536) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element and the chimeric G Protein, Gqo5, stably integrated into CHO-K1 cells.

The GeneBLAzer™ OPRK-1-Gqo5-NFAT-bla CHO-K11 cells are functionally validated for Z'-factor and EC50 concentrations using U50488. In addition, GeneBLAzer™ OPRK-1-Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_000912
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
For Use With (Application)FRET
Gene SymbolOPRK1
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer™
Quantity2 x 106 Cells
Reporter GeneBLA (Beta-Lactamase)
Target EntryOPRK1
TypeCHO-K1 Cells
Cell LineCHO-K1
System TypeGeneBLAzer™
Druggable TargetGPCR
Unit Size2 x 106 cells
Contents & Storage
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ OPRK1 Gqo5 NFAT-bla CHO-K1 cells
• GeneBLAzer™ OPRK1 Gqo5 NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ OPRK1 Gqo5 NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.