CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line
Product Image
Thermo Scientific™

CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line

Neurotrophins (NGF, BDNF, NT-3, and NT-4) and their transmembrane receptors (TrkA, TrkB, TrkC, and P75NTR) play important roles in theRead more
Have Questions?
Catalog number K1515
Price (USD)/ 1 vial
10,580.00
-
Add to cart
Price (USD)/ 1 vial
10,580.00
Add to cart
CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line
Catalog numberK1515
Price (USD)/ 1 vial
10,580.00
-
Add to cart

Neurotrophins (NGF, BDNF, NT-3, and NT-4) and their transmembrane receptors (TrkA, TrkB, TrkC, and P75NTR) play important roles in the regulation of neuronal and non-neuronal cell proliferation, differentiation, survival, and death. Neurotrophin signaling also mediates higher-order neuronal activities, such as learning, memory, and behavior. Alterations in neurotrophin levels and their receptors have been implicated in neurodegenerative disorders such as Alzheimer's disease and Huntington's disease, as well as psychiatric disorders. Neurotrophins propagate their signal through activating multiple signaling pathways. One of the signaling pathways of NT-3 (the ligand for TrkC) activates phospholipase C, releasing DAG and IP3, increasing downstream intracellular calcium, and activating protein kinase C. This in turn promotes the translocation of the transcription factor-nuclear factor of activated T-cells (NFAT)-from the cytosol into the nucleus, resulting in NFAT-dependent transcription.

The CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line was engineered by integrating the human TrkC expression plasmid into the genome of the CellSensor™ NFAT-bla CHO-K1 cell line, which is engineered to express beta-lactamase under the control of NFAT. This cell line has been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, substrate loading time, and in cryopreserved cells, and has been validated for Z'-factor and EC50 values under optimized conditions using NT-3. Additional testing information using various small-molecule inhibitors and RNAi has been performed.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
Detection MethodFluorescence
Product LineCellSensor™
Quantity1 Vial
Target EntryPKC and calcium pathway, TRKC
TypeCHO-K1 Cells
Cell LineCHO-K1
System TypeCellSensor™
Druggable TargetSignaling Pathway
Unit Size1 vial
Contents & Storage
CellSensor™ TrkC-NFAT-bla CHO-K1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.