The Tango™ NPY2R-bla U2OS cells contain the human Neuropeptide Y Receptor Y2 (NPY2R) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ NPY2R-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of Neuropeptide Y. In addition, Tango™ NPY2R-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.